Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ALG8
Entrez Gene ID
79053
Gene Name
ALG8, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1H
Chromosome Location
Chr.11: 78100942 - 78139660 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007027.2 NP_001007028.1 6 - 7 742 70
NM_024079.4 NP_076984.2 6 - 7 742 70
XM_005274247.3 XP_005274304.1 6 - 7 1503 70
XM_011545252.2 XP_011543554.1 6 - 7 752 70
XM_017018274.1 XP_016873763.1 6 - 7 1490 70
GenBank mRNA AJ224875.1 - 6 - 7 70
AK075184.1 - 6 - 7 682 70
AK075194.1 - 6 - 7 697 70
AK098614.1 - 6 - 7 742 70
AK309050.1 - 7 - 8 70
BC001133.2 - 6 - 7 725 70
KU178764.1 - 6 - 7 677 70

View Full Product Details