Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ALG8
Entrez Gene ID
79053
Gene Name
ALG8, alpha-1,3-glucosyltransferase
Gene Aliases
CDG1H
Chromosome Location
Chr.11: 78100942 - 78139660 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001007027.2 NP_001007028.1 9 - 10 1102 99
NM_024079.4 NP_076984.2 9 - 10 1102 99
XM_005274247.3 XP_005274304.1 9 - 10 1863 99
XM_011545252.2 XP_011543554.1 9 - 10 1112 99
XM_017018274.1 XP_016873763.1 9 - 10 1850 99
GenBank mRNA AJ224875.1 - 9 - 10 1056 99
AK075184.1 - 9 - 10 1042 99
AK075194.1 - 9 - 10 1057 99
AK098614.1 - 9 - 10 1102 99
BC001133.2 - 9 - 10 1085 99
KU178764.1 - 9 - 10 1037 99

View Full Product Details