Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CLINT1
Entrez Gene ID
9685
Gene Name
clathrin interactor 1
Gene Aliases
CLINT, ENTH, EPN4, EPNR
Chromosome Location
Chr.5: 157785743 - 157859175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001195555.1 NP_001182484.1 10 - 11 1580 72
NM_001195556.1 NP_001182485.1 10 - 11 1557 72
XM_011534701.2 XP_011533003.1 10 - 11 1565 72
GenBank mRNA AK092765.1 - 10 - 11 1527 72
AK300257.1 - 10 - 11 1548 72
BC013091.1 - 3 - 4 391 72
KU178487.1 - 10 - 11 1375 72

View Full Product Details