The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003502.3 | NP_003493.1 | 4 - 5 | 1508 | 78 |
NM_181050.2 | NP_851393.1 | 4 - 5 | 1508 | 78 | |
XM_017023743.1 | XP_016879232.1 | 4 - 5 | 1519 | 78 | |
XM_017023744.1 | XP_016879233.1 | 4 - 5 | 1518 | 78 | |
GenBank mRNA | AB208876.1 | - | 3 - 4 | 416 | 78 |
BC035872.1 | - | 3 - 4 | 549 | 78 | |
BC044648.1 | - | 4 - 5 | 1280 | 78 | |
KF615847.1 | - | 4 - 5 | 479 | 78 | |
KF615848.1 | - | 4 - 5 | 479 | 78 | |
KF615851.1 | - | 3 - 4 | 1367 | 78 | |
KU178413.1 | - | 3 - 4 | 1119 | 78 | |
KU178414.1 | - | 3 - 4 | 1119 | 78 | |
KU178415.1 | - | 2 - 3 | 978 | 78 |