The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003502.3 | NP_003493.1 | 5 - 6 | 1640 | 55 |
NM_181050.2 | NP_851393.1 | 5 - 6 | 1640 | 55 | |
NR_134879.1 | 4 - 5 | 1543 | 55 | ||
XM_011522682.2 | XP_011520984.1 | 5 - 6 | 1666 | 55 | |
XM_011522683.2 | XP_011520985.1 | 5 - 6 | 1665 | 55 | |
XM_011522684.2 | XP_011520986.1 | 5 - 6 | 1516 | 55 | |
XM_011522686.1 | XP_011520988.1 | 4 - 5 | 423 | 55 | |
XM_017023743.1 | XP_016879232.1 | 5 - 6 | 1651 | 55 | |
XM_017023744.1 | XP_016879233.1 | 5 - 6 | 1650 | 55 | |
XM_017023745.1 | XP_016879234.1 | 4 - 5 | 691 | 55 | |
XM_017023746.1 | XP_016879235.1 | 4 - 5 | 1267 | 55 | |
GenBank mRNA | AB208876.1 | - | 4 - 5 | 548 | 55 |
AF009674.1 | - | 5 - 6 | 1363 | 55 | |
BC035872.1 | - | 4 - 5 | 681 | 55 | |
BC044648.1 | - | 5 - 6 | 1412 | 55 | |
KF615847.1 | - | 5 - 6 | 611 | 55 | |
KF615848.1 | - | 5 - 6 | 611 | 55 | |
KF615852.1 | - | 4 - 5 | 1543 | 55 | |
KU178413.1 | - | 4 - 5 | 1251 | 55 | |
KU178414.1 | - | 4 - 5 | 1251 | 55 | |
KU178415.1 | - | 3 - 4 | 1110 | 55 |