The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003502.3 | NP_003493.1 | 6 - 7 | 2173 | 105 |
NM_181050.2 | NP_851393.1 | 6 - 7 | 2173 | 105 | |
NR_134879.1 | 5 - 6 | 2076 | 105 | ||
XM_011522682.2 | XP_011520984.1 | 6 - 7 | 2199 | 105 | |
XM_011522683.2 | XP_011520985.1 | 6 - 7 | 2198 | 105 | |
XM_011522684.2 | XP_011520986.1 | 6 - 7 | 2049 | 105 | |
XM_011522686.1 | XP_011520988.1 | 5 - 6 | 956 | 105 | |
XM_017023743.1 | XP_016879232.1 | 6 - 7 | 2184 | 105 | |
XM_017023744.1 | XP_016879233.1 | 6 - 7 | 2183 | 105 | |
XM_017023745.1 | XP_016879234.1 | 5 - 6 | 1224 | 105 | |
XM_017023746.1 | XP_016879235.1 | 5 - 6 | 1800 | 105 | |
GenBank mRNA | AB208876.1 | - | 5 - 6 | 1081 | 105 |
AF009674.1 | - | 6 - 7 | 1899 | 105 | |
BC017447.1 | - | 1 - 2 | 375 | 105 | |
BC035872.1 | - | 5 - 6 | 1214 | 105 | |
BC044648.1 | - | 6 - 7 | 1945 | 105 | |
KF615847.1 | - | 6 - 7 | 1144 | 105 | |
KF615848.1 | - | 6 - 7 | 1144 | 105 | |
KF615849.1 | - | 3 - 4 | 768 | 105 | |
KF615852.1 | - | 5 - 6 | 2076 | 105 | |
KU178413.1 | - | 5 - 6 | 1784 | 105 | |
KU178414.1 | - | 5 - 6 | 1784 | 105 | |
KU178415.1 | - | 4 - 5 | 1643 | 105 |