The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_004890.2 | NP_004881.2 | 5 - 6 | 456 | 65 |
XM_006721600.2 | XP_006721663.1 | 5 - 6 | 1081 | 65 | |
GenBank mRNA | AF047437.1 | - | 5 - 6 | 433 | 65 |
AK123065.1 | - | - | 1235 | 65 | |
AK312174.1 | - | 5 - 6 | 432 | 65 | |
BC011934.2 | - | 5 - 6 | 425 | 65 | |
BP308951.1 | - | 5 - 6 | 455 | 65 |