Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- PRH1
- Entrez Gene ID
-
5554
- Gene Name
- proline rich protein HaeIII subfamily 1
- Gene Aliases
-
Db-s, PA, PIF-S, PRH2, Pr1/Pr2
- Chromosome Location
-
Chr.12: 10880961 - 11171623 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BU543118.1 |
- |
2 - 3 |
361 |
107 |
View Full Product Details