The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206838.1 | NP_001193767.1 | 17 - 18 | 3092 | 90 |
NM_001206839.1 | NP_001193768.1 | 16 - 17 | 3071 | 90 | |
NM_002851.2 | NP_002842.2 | 17 - 18 | 5672 | 90 | |
XM_005250519.1 | XP_005250576.1 | 16 - 17 | 5651 | 90 | |
XM_017012477.1 | XP_016867966.1 | 18 - 19 | 5719 | 90 | |
GenBank mRNA | AK294062.1 | - | 6 - 7 | 926 | 90 |
M93426.1 | - | 17 - 18 | 5421 | 90 | |
X54135.1 | - | 6 - 7 | 840 | 90 |