The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206838.1 | NP_001193767.1 | 4 - 5 | 853 | 108 |
NM_001206839.1 | NP_001193768.1 | 4 - 5 | 853 | 108 | |
NM_002851.2 | NP_002842.2 | 4 - 5 | 853 | 108 | |
XM_005250519.1 | XP_005250576.1 | 4 - 5 | 853 | 108 | |
XM_017012477.1 | XP_016867966.1 | 5 - 6 | 900 | 108 | |
GenBank mRNA | M93426.1 | - | 4 - 5 | 605 | 108 |