Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CPE
Entrez Gene ID
1363
Gene Name
carboxypeptidase E
Gene Aliases
CPH
Chromosome Location
Chr.4: 165378942 - 165498547 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001873.3 NP_001864.1 1 - 2 585 84
GenBank mRNA AK090962.1 - 2 - 3 369 84
AK127165.1 - 2 - 3 302 84
AK290996.1 - 1 - 2 444 84
AK294175.1 - 2 - 3 402 84
BC033866.1 - 1 - 2 323 84
BC053612.1 - 1 - 2 437 84
X51405.1 - 1 - 2 595 84

View Full Product Details