The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022478.3 | NP_071923.2 | 11 - 12 | 1978 | 123 |
NM_144985.3 | NP_659422.2 | 10 - 11 | 1864 | 123 | |
GenBank mRNA | AK025342.1 | - | 3 - 4 | 282 | 123 |
AK289576.1 | - | 10 - 11 | 1864 | 123 | |
AL137477.1 | - | 2 - 3 | 437 | 123 | |
AY260900.1 | - | 9 - 10 | 1697 | 123 | |
AY260901.1 | - | 10 - 11 | 1811 | 123 | |
AY358199.1 | - | 10 - 11 | 1820 | 123 | |
BX248303.1 | - | 6 - 7 | 888 | 123 |