The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025235.3 | NP_079511.1 | 23 - 24 | 3284 | 85 |
XM_005270185.4 | XP_005270242.2 | 23 - 24 | 3324 | 85 | |
XM_011540213.1 | XP_011538515.1 | 23 - 24 | 3315 | 85 | |
XM_011540215.2 | XP_011538517.1 | 15 - 16 | 3509 | 85 | |
XM_017016696.1 | XP_016872185.1 | 23 - 24 | 3261 | 85 | |
XM_017016697.1 | XP_016872186.1 | 23 - 24 | 3277 | 85 | |
XM_017016698.1 | XP_016872187.1 | 23 - 24 | 2988 | 85 | |
XM_017016699.1 | XP_016872188.1 | 23 - 24 | 3268 | 85 | |
XM_017016700.1 | XP_016872189.1 | 23 - 24 | 2874 | 85 | |
GenBank mRNA | AF264912.1 | - | 23 - 24 | 2987 | 85 |
AF305081.1 | - | 23 - 24 | 3286 | 85 | |
AF309033.1 | - | 23 - 24 | 3238 | 85 | |
AF329696.1 | - | 23 - 24 | 3126 | 85 | |
AF342982.1 | - | 23 - 24 | 2987 | 85 | |
AF438201.1 | - | 23 - 24 | 3366 | 85 | |
AK023746.1 | - | 5 - 6 | 478 | 85 | |
AK314612.1 | - | 23 - 24 | 3252 | 85 | |
AL833676.1 | - | 9 - 10 | 1322 | 85 |