The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025235.3 | NP_079511.1 | 6 - 7 | 1020 | 81 |
XM_005270185.4 | XP_005270242.2 | 6 - 7 | 1051 | 81 | |
XM_011540213.1 | XP_011538515.1 | 6 - 7 | 1051 | 81 | |
XM_017016696.1 | XP_016872185.1 | 6 - 7 | 988 | 81 | |
XM_017016697.1 | XP_016872186.1 | 6 - 7 | 1004 | 81 | |
XM_017016698.1 | XP_016872187.1 | 6 - 7 | 715 | 81 | |
XM_017016699.1 | XP_016872188.1 | 6 - 7 | 1004 | 81 | |
XM_017016700.1 | XP_016872189.1 | 6 - 7 | 601 | 81 | |
XM_017016701.1 | XP_016872190.1 | 6 - 7 | 1051 | 81 | |
GenBank mRNA | AF264912.1 | - | 6 - 7 | 723 | 81 |
AF305081.1 | - | 6 - 7 | 1025 | 81 | |
AF309033.1 | - | 6 - 7 | 974 | 81 | |
AF329696.1 | - | 6 - 7 | 862 | 81 | |
AF342982.1 | - | 6 - 7 | 723 | 81 | |
AF438201.1 | - | 6 - 7 | 1102 | 81 | |
AK314612.1 | - | 6 - 7 | 988 | 81 |