Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD2AP
Entrez Gene ID
23607
Gene Name
CD2 associated protein
Gene Aliases
CMS
Chromosome Location
Chr.6: 47477746 - 47627263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012120.2 NP_036252.1 5 - 6 990 105
XM_005248976.1 XP_005249033.1 5 - 6 1033 105
XM_011514449.2 XP_011512751.1 4 - 5 612 105
XM_017010641.1 XP_016866130.1 5 - 6 1033 105
GenBank mRNA AF146277.1 - 5 - 6 584 105
AF164377.1 - 4 - 5 534 105
BC069444.1 - 5 - 6 580 105

View Full Product Details