Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD2AP
Entrez Gene ID
23607
Gene Name
CD2 associated protein
Gene Aliases
CMS
Chromosome Location
Chr.6: 47477746 - 47627263 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012120.2 NP_036252.1 8 - 9 1356 96
XM_005248976.1 XP_005249033.1 8 - 9 1387 96
XM_011514449.2 XP_011512751.1 7 - 8 978 96
XM_017010641.1 XP_016866130.1 8 - 9 1399 96
GenBank mRNA AF146277.1 - 8 - 9 950 96
AF164377.1 - 7 - 8 900 96
BC069444.1 - 8 - 9 946 96

View Full Product Details