Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXJ3
Entrez Gene ID
22887
Gene Name
forkhead box J3
Gene Aliases
-
Chromosome Location
Chr.1: 42176539 - 42335877 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198850.1 NP_001185779.1 4 - 5 716 127
NM_001198851.1 NP_001185780.1 6 - 7 759 127
NM_001198852.1 NP_001185781.1 4 - 5 621 127
NM_014947.4 NP_055762.3 4 - 5 638 127
XM_005270632.4 XP_005270689.1 3 - 4 526 127
XM_006710458.3 XP_006710521.1 4 - 5 745 127
XM_006710459.3 XP_006710522.1 3 - 4 527 127
XM_011541026.2 XP_011539328.1 5 - 6 690 127
XM_017000693.1 XP_016856182.1 4 - 5 765 127
GenBank mRNA AB028964.1 - 6 - 7 759 127
AK027075.1 - 4 - 5 716 127
BC014182.2 - 4 - 5 621 127
BC151828.1 - 4 - 5 607 127
BC152441.1 - 6 - 7 759 127

View Full Product Details