Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FOXJ3
Entrez Gene ID
22887
Gene Name
forkhead box J3
Gene Aliases
-
Chromosome Location
Chr.1: 42176539 - 42335877 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001198850.1 NP_001185779.1 7 - 8 1032 81
NM_001198851.1 NP_001185780.1 9 - 10 1075 81
NM_001198852.1 NP_001185781.1 6 - 7 835 81
NM_014947.4 NP_055762.3 7 - 8 954 81
XM_005270632.4 XP_005270689.1 6 - 7 842 81
XM_006710458.3 XP_006710521.1 7 - 8 1061 81
XM_006710459.3 XP_006710522.1 5 - 6 741 81
XM_011541026.2 XP_011539328.1 8 - 9 1006 81
XM_017000693.1 XP_016856182.1 6 - 7 979 81
XM_017000694.1 XP_016856183.1 5 - 6 718 81
XM_017000695.1 XP_016856184.1 6 - 7 604 81
GenBank mRNA AB028964.1 - 9 - 10 1075 81
AK027075.1 - 7 - 8 1032 81
BC014182.2 - 6 - 7 835 81
BC151828.1 - 7 - 8 923 81
BC152441.1 - 9 - 10 1075 81

View Full Product Details