The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012433.3 | NP_036565.2 | 21 - 22 | 3226 | 137 |
GenBank mRNA | AF054284.1 | - | 21 - 22 | 3132 | 137 |
AF070540.1 | - | 1 - 2 | 104 | 137 |