The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012433.3 | NP_036565.2 | 23 - 24 | 3626 | 134 |
GenBank mRNA | AF054284.1 | - | 23 - 24 | 3532 | 134 |
AF070540.1 | - | 3 - 4 | 504 | 134 |