The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_033229.2 | NP_150232.2 | 2 - 3 | 951 | 118 |
XM_011514987.1 | XP_011513289.1 | 3 - 4 | 190 | 118 | |
XM_011514988.2 | XP_011513290.1 | 2 - 3 | 472 | 118 | |
GenBank mRNA | AF220132.1 | - | 2 - 3 | 472 | 118 |
AK290280.1 | - | 2 - 3 | 657 | 118 | |
AK291853.1 | - | 2 - 3 | 609 | 118 | |
BC038585.1 | - | 2 - 3 | 552 | 118 | |
U34249.1 | - | 2 - 3 | 965 | 118 |