The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014362.3 | NP_055177.2 | 4 - 5 | 601 | 84 |
NM_198047.2 | NP_932164.1 | 4 - 5 | 601 | 84 | |
XM_011510953.1 | XP_011509255.1 | 4 - 5 | 550 | 84 | |
GenBank mRNA | AK223023.1 | - | 4 - 5 | 325 | 84 |
AK308042.1 | - | 4 - 5 | 325 | 84 | |
AK313651.1 | - | 4 - 5 | 331 | 84 | |
BC005190.1 | - | 4 - 5 | 378 | 84 | |
BC067822.1 | - | 4 - 5 | 330 | 84 | |
BI764401.1 | - | 4 - 5 | 411 | 84 | |
U66669.1 | - | 4 - 5 | 354 | 84 |