The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014362.3 | NP_055177.2 | 7 - 8 | 811 | 85 |
NM_198047.2 | NP_932164.1 | 7 - 8 | 811 | 85 | |
XM_011510953.1 | XP_011509255.1 | 7 - 8 | 760 | 85 | |
XM_011510954.1 | XP_011509256.1 | 2 - 3 | 211 | 85 | |
GenBank mRNA | AK222979.1 | - | 7 - 8 | 535 | 85 |
AK223023.1 | - | 7 - 8 | 535 | 85 | |
AK308042.1 | - | 7 - 8 | 535 | 85 | |
AK313651.1 | - | 7 - 8 | 541 | 85 | |
BC005190.1 | - | 7 - 8 | 588 | 85 | |
BC067822.1 | - | 7 - 8 | 540 | 85 |