The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030666.3 | NP_109591.1 | 3 - 4 | 549 | 133 |
NR_073111.1 | 4 - 5 | 681 | 133 | ||
NR_073112.1 | 3 - 4 | 549 | 133 | ||
XM_011514333.1 | XP_011512635.1 | 4 - 5 | 772 | 133 | |
XM_011514334.2 | XP_011512636.1 | 3 - 4 | 683 | 133 | |
XM_011514335.2 | XP_011512637.1 | 3 - 4 | 538 | 133 | |
GenBank mRNA | AK223370.1 | - | 3 - 4 | 362 | 133 |
AK291327.1 | - | 3 - 4 | 362 | 133 | |
AK298044.1 | - | 4 - 5 | 492 | 133 | |
AK304641.1 | - | 3 - 4 | 394 | 133 | |
AK309765.1 | - | 4 - 5 | 683 | 133 | |
AK312973.1 | - | 3 - 4 | 360 | 133 | |
BC009015.1 | - | 3 - 4 | 347 | 133 | |
BT006928.1 | - | 2 - 3 | 303 | 133 | |
BU942473.1 | - | 3 - 4 | 364 | 133 | |
EU794624.1 | - | 2 - 3 | 303 | 133 | |
GQ472232.1 | - | 3 - 4 | 347 | 133 | |
M93056.1 | - | 3 - 4 | 351 | 133 |