The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030666.3 | NP_109591.1 | 5 - 6 | 818 | 102 |
NR_073111.1 | 6 - 7 | 950 | 102 | ||
XM_011514333.1 | XP_011512635.1 | 6 - 7 | 1041 | 102 | |
XM_011514334.2 | XP_011512636.1 | 5 - 6 | 952 | 102 | |
XM_011514335.2 | XP_011512637.1 | 5 - 6 | 807 | 102 | |
GenBank mRNA | AK223370.1 | - | 5 - 6 | 631 | 102 |
AK291327.1 | - | 5 - 6 | 631 | 102 | |
AK298044.1 | - | 6 - 7 | 760 | 102 | |
AK304641.1 | - | 5 - 6 | 663 | 102 | |
AK312973.1 | - | 5 - 6 | 629 | 102 | |
BC009015.1 | - | 5 - 6 | 616 | 102 | |
BT006928.1 | - | 4 - 5 | 572 | 102 | |
EU794624.1 | - | 4 - 5 | 572 | 102 | |
GQ472232.1 | - | 5 - 6 | 616 | 102 | |
M93056.1 | - | 5 - 6 | 620 | 102 |