The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001317004.1 | NP_001303933.1 | 1 - 2 | 77 | 124 |
XM_011510623.2 | XP_011508925.1 | 2 - 3 | 164 | 124 | |
GenBank mRNA | AF435957.1 | - | 1 - 2 | 124 |