Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CFH
Entrez Gene ID
3075
Gene Name
complement factor H
Gene Aliases
AHUS1, AMBP1, ARMD4, ARMS1, CFHL3, FH, FHL1, HF, HF1, HF2, HUS
Chromosome Location
Chr.1: 196651878 - 196747504 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000186.3 NP_000177.2 16 - 17 2829 67
GenBank mRNA AK226113.1 - 11 - 12 4615 67
AK291395.1 - 16 - 17 2668 67
BC142699.1 - 16 - 17 2641 67
M17517.1 - 5 - 6 868 67
Y00716.1 - 16 - 17 2662 67

View Full Product Details