The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015456.4 | NP_056271.3 | 1 - 2 | 310 | 51 |
GenBank mRNA | AF464935.1 | - | 1 - 2 | 115 | 51 |
AK091056.1 | - | 1 - 2 | 303 | 51 | |
BC019029.1 | - | 1 - 2 | 269 | 51 | |
DA210427.1 | - | 1 - 2 | 304 | 51 |