The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015456.4 | NP_056271.3 | 7 - 8 | 1207 | 145 |
GenBank mRNA | AF464935.1 | - | 7 - 8 | 1012 | 145 |
AK022651.1 | - | 5 - 6 | 703 | 145 | |
AK091056.1 | - | 7 - 8 | 1200 | 145 | |
AL050280.1 | - | 4 - 5 | 454 | 145 | |
BC011892.1 | - | 6 - 7 | 916 | 145 | |
BC019029.1 | - | 7 - 8 | 1166 | 145 |