The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_032588.3 | NP_115977.2 | 7 - 8 | 1112 | 84 |
XM_017002559.1 | XP_016858048.1 | 7 - 8 | 1112 | 84 | |
GenBank mRNA | AF353673.1 | - | 7 - 8 | 1091 | 84 |
AJ276484.1 | - | 7 - 8 | 1095 | 84 | |
AK056942.1 | - | 7 - 8 | 1108 | 84 | |
AK297820.1 | - | 7 - 8 | 1024 | 84 | |
BC080529.1 | - | 7 - 8 | 1090 | 84 |