Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SATB1
Entrez Gene ID
6304
Gene Name
SATB homeobox 1
Gene Aliases
-
Chromosome Location
Chr.3: 18345388 - 18445588 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001131010.3 NP_001124482.1 7 - 8 1437 109
NM_001195470.2 NP_001182399.1 7 - 8 2939 109
NM_001322871.1 NP_001309800.1 7 - 8 1485 109
NM_001322872.1 NP_001309801.1 8 - 9 1485 109
NM_001322873.1 NP_001309802.1 7 - 8 1485 109
NM_001322874.1 NP_001309803.1 7 - 8 1299 109
NM_001322875.1 NP_001309804.1 7 - 8 1297 109
NM_001322876.1 NP_001309805.1 6 - 7 1250 109
NM_002971.5 NP_002962.1 7 - 8 2939 109
XM_011533988.2 XP_011532290.1 7 - 8 1399 109
XM_011533989.2 XP_011532291.1 7 - 8 1299 109
XM_011533990.2 XP_011532292.1 7 - 8 2939 109
GenBank mRNA AB209761.1 - 7 - 8 2211 109
AK127242.1 - 7 - 8 2944 109
BC001744.1 - 7 - 8 1376 109
M97287.1 - 7 - 8 1418 109

View Full Product Details