Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
THUMPD3
Entrez Gene ID
25917
Gene Name
THUMP domain containing 3
Gene Aliases
-
Chromosome Location
Chr.3: 9362842 - 9386791 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114092.1 NP_001107564.1 8 - 9 1387 103
NM_015453.2 NP_056268.2 8 - 9 1592 103
XM_005265021.4 XP_005265078.1 8 - 9 1949 103
XM_005265022.4 XP_005265079.1 8 - 9 2152 103
XM_005265023.3 XP_005265080.1 8 - 9 1535 103
XM_005265024.4 XP_005265081.1 8 - 9 1535 103
XM_006713086.3 XP_006713149.1 8 - 9 2191 103
XM_011533570.2 XP_011531872.1 8 - 9 1655 103
XM_011533571.2 XP_011531873.1 6 - 7 1406 103
XM_017006113.1 XP_016861602.1 7 - 8 1833 103
XM_017006114.1 XP_016861603.1 7 - 8 2075 103
XM_017006115.1 XP_016861604.1 7 - 8 1476 103
XM_017006116.1 XP_016861605.1 7 - 8 1462 103
XM_017006117.1 XP_016861606.1 6 - 7 1763 103
XM_017006118.1 XP_016861607.1 6 - 7 2005 103
XM_017006119.1 XP_016861608.1 6 - 7 1392 103
GenBank mRNA AK001680.1 - 8 - 9 1351 103
AL117483.1 - 5 - 6 810 103
BC001622.1 - 8 - 9 103
BC010421.1 - 8 - 9 1923 103

View Full Product Details