Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
MTX2
Entrez Gene ID
10651
Gene Name
metaxin 2
Gene Aliases
-
Chromosome Location
Chr.2: 176269395 - 176338025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001319097.1 NP_001306026.1 1 - 2 281 85
NM_001319098.1 NP_001306027.1 1 - 2 281 85
NM_006554.4 NP_006545.1 1 - 2 281 85
XM_011510497.2 XP_011508799.1 1 - 2 253 85
GenBank mRNA AF053551.1 - 1 - 2 254 85
AK001287.1 - 1 - 2 233 85
AK289359.1 - 1 - 2 232 85
BC067831.1 - 1 - 2 260 85
BC088359.1 - 1 - 2 216 85
BU165147.1 - 1 - 2 212 85
CX165048.1 - 1 - 2 174 85

View Full Product Details