Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
MTX2
Entrez Gene ID
10651
Gene Name
metaxin 2
Gene Aliases
-
Chromosome Location
Chr.2: 176269395 - 176338025 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006635.2 NP_001006636.1 7 - 8 818 118
NM_001319097.1 NP_001306026.1 6 - 7 614 118
NM_001319098.1 NP_001306027.1 6 - 7 614 118
NM_006554.4 NP_006545.1 6 - 7 614 118
XM_011510497.2 XP_011508799.1 7 - 8 711 118
GenBank mRNA AF053551.1 - 6 - 7 587 118
AK001287.1 - 6 - 7 566 118
AK289359.1 - 6 - 7 565 118
AK303564.1 - 7 - 8 743 118
AK315918.1 - 7 - 8 754 118
BC017271.2 - 7 - 8 748 118
BC067831.1 - 6 - 7 593 118
BC088359.1 - 6 - 7 549 118
BU165147.1 - 6 - 7 545 118
CX165048.1 - 6 - 7 507 118

View Full Product Details