Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
STAM2
Entrez Gene ID
10254
Gene Name
signal transducing adaptor molecule 2
Gene Aliases
Hbp, STAM2A, STAM2B
Chromosome Location
Chr.2: 152116801 - 152175992 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF042274.1 - 10 - 11 1081 140

View Full Product Details