The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005843.5 | NP_005834.4 | 1 - 2 | 388 | 145 |
GenBank mRNA | AF042273.1 | - | 1 - 2 | 147 | 145 |
AF042274.1 | - | 1 - 2 | 147 | 145 | |
AK292265.1 | - | 1 - 2 | 83 | 145 | |
AK292847.1 | - | 1 - 2 | 125 | 145 | |
AK310863.1 | - | 1 - 2 | 125 | 145 | |
BC028740.2 | - | 1 - 2 | 388 | 145 | |
BI548350.1 | - | 1 - 2 | 393 | 145 |