The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005843.5 | NP_005834.4 | 4 - 5 | 652 | 100 |
GenBank mRNA | AF042273.1 | - | 4 - 5 | 411 | 100 |
AF042274.1 | - | 4 - 5 | 411 | 100 | |
AK292265.1 | - | 4 - 5 | 347 | 100 | |
AK292847.1 | - | 4 - 5 | 389 | 100 | |
AK310863.1 | - | 4 - 5 | 389 | 100 | |
AL133600.1 | - | 4 - 5 | 414 | 100 | |
BC028740.2 | - | 4 - 5 | 652 | 100 |