Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C2orf40
Entrez Gene ID
84417
Gene Name
chromosome 2 open reading frame 40
Gene Aliases
ECRG4
Chromosome Location
Chr.2: 106063290 - 106078155 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032411.2 NP_115787.1 3 - 4 395 110
XM_006712799.3 XP_006712862.1 4 - 5 414 110
XM_011511993.2 XP_011510295.1 3 - 4 332 110
XM_017005106.1 XP_016860595.1 3 - 4 314 110
GenBank mRNA AF325503.1 - 3 - 4 395 110
AY358601.1 - 3 - 4 110
BC021742.1 - 3 - 4 399 110

View Full Product Details