The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000925.3 | NP_000916.2 | 4 - 5 | 316 | 92 |
NM_001173468.1 | NP_001166939.1 | 4 - 5 | 316 | 92 | |
NM_001315536.1 | NP_001302465.1 | 3 - 4 | 262 | 92 | |
NR_033384.1 | 3 - 4 | 386 | 92 | ||
GenBank mRNA | AK225569.1 | - | 4 - 5 | 291 | 92 |
AK225621.1 | - | 4 - 5 | 296 | 92 | |
AK293153.1 | - | 4 - 5 | 282 | 92 | |
AK311514.1 | - | 4 - 5 | 291 | 92 | |
AK313022.1 | - | 4 - 5 | 291 | 92 | |
AL117618.1 | - | 3 - 4 | 227 | 92 | |
BC000439.2 | - | 4 - 5 | 283 | 92 | |
BC001924.1 | - | 4 - 5 | 283 | 92 | |
CR541911.1 | - | 4 - 5 | 273 | 92 | |
DA965753.1 | - | 4 - 5 | 316 | 92 | |
DB039833.1 | - | 3 - 4 | 386 | 92 | |
J03576.1 | - | 4 - 5 | 291 | 92 | |
M34055.1 | - | 4 - 5 | 278 | 92 | |
M34056.1 | - | 3 - 4 | 201 | 92 | |
M34479.1 | - | 4 - 5 | 273 | 92 | |
M54788.1 | - | 4 - 5 | 278 | 92 | |
X57778.1 | - | 4 - 5 | 292 | 92 |