The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006421.4 | NP_006412.2 | 2 - 3 | 547 | 72 |
XM_005251134.4 | XP_005251191.1 | 3 - 4 | 526 | 72 | |
XM_005251135.3 | XP_005251192.1 | 2 - 3 | 797 | 72 | |
XM_005251136.3 | XP_005251193.1 | 2 - 3 | 797 | 72 | |
GenBank mRNA | AF084520.1 | - | 2 - 3 | 157 | 72 |
AF111162.1 | - | 2 - 3 | 298 | 72 | |
AI523655.1 | - | 3 - 4 | 374 | 72 |