The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006421.4 | NP_006412.2 | 37 - 38 | 5660 | 112 |
XM_005251134.4 | XP_005251191.1 | 38 - 39 | 5639 | 112 | |
XM_005251135.3 | XP_005251192.1 | 37 - 38 | 5892 | 112 | |
XM_005251136.3 | XP_005251193.1 | 37 - 38 | 5910 | 112 | |
XM_011517442.2 | XP_011515744.1 | 38 - 39 | 5946 | 112 | |
GenBank mRNA | AB209324.1 | - | 27 - 28 | 3705 | 112 |
AF084520.1 | - | 37 - 38 | 5270 | 112 | |
AF111162.1 | - | 37 - 38 | 5411 | 112 | |
AK001788.1 | - | 10 - 11 | 1304 | 112 | |
AK022557.1 | - | 13 - 14 | 1759 | 112 | |
AK025637.1 | - | 18 - 19 | 2259 | 112 | |
AK123660.1 | - | 1 - 2 | 342 | 112 | |
AK127799.1 | - | 4 - 5 | 2858 | 112 | |
AL117446.1 | - | 3 - 4 | 363 | 112 |