The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256358.1 | NP_001243287.1 | 5 - 6 | 945 | 86 |
NM_005975.3 | NP_005966.1 | 6 - 7 | 1067 | 86 | |
GenBank mRNA | AK124082.1 | - | 2 - 3 | 1027 | 86 |
AK301364.1 | - | 5 - 6 | 945 | 86 | |
AK315232.1 | - | 6 - 7 | 1049 | 86 | |
BC035843.1 | - | 6 - 7 | 1051 | 86 | |
X78549.1 | - | 6 - 7 | 1039 | 86 |