Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PAPOLG
Entrez Gene ID
64895
Gene Name
poly(A) polymerase gamma
Gene Aliases
-
Chromosome Location
Chr.2: 60756230 - 60802086 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022894.3 NP_075045.2 10 - 11 1155 94
XM_005264500.3 XP_005264557.1 10 - 11 1155 94
XM_005264501.1 XP_005264558.1 10 - 11 973 94
XM_011533041.1 XP_011531343.1 9 - 10 861 94
XM_017004723.1 XP_016860212.1 9 - 10 1017 94
XM_017004724.1 XP_016860213.1 9 - 10 1017 94
GenBank mRNA AF312211.1 - 10 - 11 1137 94
AJ308101.1 - 10 - 11 906 94
AK021867.1 - 10 - 11 1091 94
AK023544.1 - 9 - 10 904 94
AK314663.1 - 10 - 11 1119 94
BC016738.1 - 10 - 11 1126 94
BC110906.1 - 10 - 11 1094 94
BC111701.1 - 10 - 11 1029 94
BC113747.1 - 10 - 11 1029 94
BX647824.1 - 9 - 10 875 94

View Full Product Details