The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022894.3 | NP_075045.2 | 8 - 9 | 944 | 76 |
XM_005264500.3 | XP_005264557.1 | 8 - 9 | 944 | 76 | |
XM_005264501.1 | XP_005264558.1 | 8 - 9 | 762 | 76 | |
XM_011533041.1 | XP_011531343.1 | 7 - 8 | 650 | 76 | |
XM_017004723.1 | XP_016860212.1 | 7 - 8 | 806 | 76 | |
XM_017004724.1 | XP_016860213.1 | 7 - 8 | 806 | 76 | |
GenBank mRNA | AF312211.1 | - | 8 - 9 | 926 | 76 |
AJ308101.1 | - | 8 - 9 | 695 | 76 | |
AK021867.1 | - | 8 - 9 | 880 | 76 | |
AK023544.1 | - | 7 - 8 | 693 | 76 | |
AK314663.1 | - | 8 - 9 | 908 | 76 | |
AY029162.1 | - | 8 - 9 | 704 | 76 | |
BC016738.1 | - | 8 - 9 | 915 | 76 | |
BC110906.1 | - | 8 - 9 | 883 | 76 | |
BC111701.1 | - | 8 - 9 | 818 | 76 | |
BC113747.1 | - | 8 - 9 | 818 | 76 | |
BX647824.1 | - | 7 - 8 | 664 | 76 |