The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006729.4 | NP_006720.1 | 13 - 14 | 1842 | 133 |
NM_007309.3 | NP_009293.1 | 13 - 14 | 1842 | 133 | |
GenBank mRNA | AK002029.1 | - | 1 - 2 | 114 | 133 |
AK291272.1 | - | 13 - 14 | 1724 | 133 | |
BC117414.1 | - | 14 - 15 | 1509 | 133 | |
Y15908.1 | - | 13 - 14 | 1796 | 133 | |
Y15909.1 | - | 13 - 14 | 1796 | 133 |