The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001139488.1 | NP_001132960.1 | 3 - 4 | 784 | 91 |
NM_015376.2 | NP_056191.1 | 2 - 3 | 342 | 91 | |
NM_170672.2 | NP_733772.1 | 4 - 5 | 697 | 91 | |
XM_005264246.3 | XP_005264303.1 | 3 - 4 | 1124 | 91 | |
XM_011532746.2 | XP_011531048.1 | 4 - 5 | 571 | 91 | |
XM_011532747.2 | XP_011531049.1 | 3 - 4 | 586 | 91 | |
XM_011532748.2 | XP_011531050.2 | 3 - 4 | 432 | 91 | |
XM_017003759.1 | XP_016859248.1 | 5 - 6 | 2047 | 91 | |
XM_017003760.1 | XP_016859249.1 | 4 - 5 | 2519 | 91 | |
XM_017003761.1 | XP_016859250.1 | 3 - 4 | 449 | 91 | |
XM_017003762.1 | XP_016859251.1 | 4 - 5 | 698 | 91 | |
XM_017003763.1 | XP_016859252.1 | 3 - 4 | 428 | 91 | |
GenBank mRNA | AB020653.1 | - | 2 - 3 | 342 | 91 |
AK292983.1 | - | 4 - 5 | 608 | 91 | |
BC027849.1 | - | 4 - 5 | 697 | 91 | |
BX647990.1 | - | 3 - 4 | 392 | 91 |