The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001288653.1 | NP_001275582.1 | 5 - 6 | 1244 | 125 |
NM_004859.3 | NP_004850.1 | 5 - 6 | 1232 | 125 | |
XM_005257012.3 | XP_005257069.1 | 5 - 6 | 1044 | 125 | |
GenBank mRNA | AK127134.1 | - | 5 - 6 | 1009 | 125 |
BC051800.2 | - | 5 - 6 | 1001 | 125 | |
BC054489.1 | - | 5 - 6 | 1016 | 125 | |
BX640615.1 | - | 5 - 6 | 1016 | 125 | |
D21260.1 | - | 5 - 6 | 961 | 125 |