Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
QRICH2
Entrez Gene ID
84074
Gene Name
glutamine rich 2
Gene Aliases
-
Chromosome Location
Chr.17: 76271296 - 76308690 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_032134.2 NP_115510.1 4 - 5 3392 77
XM_005257728.3 XP_005257785.1 4 - 5 4402 77
XM_011525344.2 XP_011523646.1 2 - 3 3075 77
XM_017025206.1 XP_016880695.1 4 - 5 4402 77
XM_017025207.1 XP_016880696.1 3 - 4 4342 77
GenBank mRNA AK058102.1 - 1 - 2 236 77
AL136774.1 - 4 - 5 3392 77

View Full Product Details