Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SERPINB1
Entrez Gene ID
1992
Gene Name
serpin family B member 1
Gene Aliases
EI, ELANH2, HEL-S-27, HEL57, LEI, M/NEI, MNEI, PI-2, PI2
Chromosome Location
Chr.6: 2832332 - 2842049 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011514333.1 XP_011512635.1 2 - 3 461 86

View Full Product Details