The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022894.3 | NP_075045.2 | 20 - 21 | 2305 | 139 |
XM_005264501.1 | XP_005264558.1 | 20 - 21 | 2123 | 139 | |
XM_011533041.1 | XP_011531343.1 | 19 - 20 | 2011 | 139 | |
XM_017004723.1 | XP_016860212.1 | 19 - 20 | 2167 | 139 | |
GenBank mRNA | AF312211.1 | - | 20 - 21 | 2287 | 139 |
AJ308101.1 | - | 20 - 21 | 2056 | 139 | |
AK023034.1 | - | 9 - 10 | 1007 | 139 | |
AK024249.1 | - | 8 - 9 | 922 | 139 | |
AK314663.1 | - | 20 - 21 | 2269 | 139 | |
AY029162.1 | - | 20 - 21 | 2065 | 139 | |
BC111701.1 | - | 20 - 21 | 2179 | 139 | |
BC113747.1 | - | 20 - 21 | 2179 | 139 |